missing translation for 'onlineSavingsMsg'
Learn More

Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11373807
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373807 5 g 5g
11363807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11373807 Supplier Thermo Scientific Alfa Aesar Supplier No. 035660.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.

Zinc arsenide is used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-40-5
Molecular Formula As2Zn3
Molecular Weight (g/mol) 345.98
MDL Number MFCD00049629
InChI Key RHKSESDHCKYTHI-UHFFFAOYSA-N
Synonym zinc arsenide, trizincarsa-1,4-diene, zinc arsenide, pieces trace metals basis
PubChem CID 25147458
IUPAC Name bis(zincio-λ²-arsanyl) zinc
SMILES [Zn].[Zn]=[As].[Zn]=[As]

Specifications

Melting Point 1015°C
Physical Form Pieces
Quantity 5 g
Packaging Glass bottle
Specific Gravity 5.528
Density 5.528
UN Number UN1557
Solubility Information Insoluble in water. Soluble in dilute sulfuric acid.
Formula Weight 345.95
Percent Purity 99.999%
Chemical Name or Material Zinc arsenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.