missing translation for 'onlineSavingsMsg'
Learn More

Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050215
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
12044-55-2
As2Zn
215.223
MFCD00151429
KXUSEUKXCIKTBF-UHFFFAOYSA-N
zinc arsenide znas2, 1,2-diarsa-3-zincacycloprop-1-ene
102601572
$l^{2}-arsanylidenearsenic;zinc
[Zn].[As]=[As]
This item is not returnable. View return policy
12044-55-2
As2Zn
215.223
MFCD00151429
KXUSEUKXCIKTBF-UHFFFAOYSA-N
zinc arsenide znas2, 1,2-diarsa-3-zincacycloprop-1-ene
102601572
$l^{2}-arsanylidenearsenic;zinc
[Zn].[As]=[As]

Zinc arsenide is often used as a component in the manufacture of semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Lump
Quantity 2 g
UN Number UN1557
Solubility Information It is insoluble in water.
Formula Weight 215.21
Percent Purity 99.999%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc arsenide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'