missing translation for 'onlineSavingsMsg'
Learn More

Zinc acrylate

Product Code. 11368928
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368928 1 kg 1kg
11358928 50 g 50g
11378928 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368928 Supplier Thermo Scientific Alfa Aesar Supplier No. 041780.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14643-87-9 | C6H6O4Zn | 207.49 g/mol

The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14643-87-9
Molecular Formula C6H6O4Zn
Molecular Weight (g/mol) 207.49
MDL Number MFCD00067208
InChI Key XKMZOFXGLBYJLS-UHFFFAOYSA-L
Synonym zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559
PubChem CID 159747
IUPAC Name zinc;prop-2-enoate
SMILES [Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C

Specifications

Melting Point ∼250°C
Density 1.6 g/mL
Quantity 1 kg
UN Number UN3077
Solubility Information Soluble completely in acrylic acid and water (as a salt). Insoluble as a polymer.
Formula Weight 207.5
Physical Form Powder
Chemical Name or Material Zinc acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.