missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, TRC

Product Code. 30134514
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134514 1 g 1g
30009110 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134514 Supplier Toronto Research Chemicals Supplier No. TRCZ4381001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-34-6
Molecular Formula C4H6O4Zn
Molecular Weight (g/mol) 183.47
Synonym cetic acid, zinc salt (8CI,9CI), Zinc acetate (6CI,7CI), Galzin, Siltex CL 4, Zinc diacetate, Zinc(II) acetate
IUPAC Name zinc;diacetate
SMILES [Zn+2].CC(=O)[O-].CC(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C2H4O2.Zn/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
Formula Weight 181.9558
Chemical Name or Material Zinc Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.