missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, Puriss., meets analytical spec. of DAB, E 650, 99-102%, Solstice

Product Code. 15632730
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632730 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15632730 Supplier Honeywell-Fluka Supplier No. 2504425KG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 5970-45-6
CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 99 to 102%
Molecular Formula C4H10O6Zn
Linear Formula CH3COONa · 3H2O
MDL Number MFCD00071557
Beilstein 3732513
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
IUPAC Name zinc;diacetate;dihydrate
Molecular Weight (g/mol) 219.498
PubChem CID 2724192
Formula Weight 219.51
Grade puriss.
Packaging Paper / Poly Bag
Density 1.750 g/cm3
Chemical Name or Material Zinc acetate dihydrate, Puriss., meets analytical specification of DAB, E 650, 99-102%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.