missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Acetate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Formula Weight | 219.51 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.74kg |
| Chemical Name or Material | Zinc Acetate, Dihydrate |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction