Learn More
Zinc, 99.999% (trace metal basis), powder, -40 mesh, Thermo Scientific Chemicals
£112.00 - £381.00
Chemical Identifiers
CAS | 7440-66-6 |
---|---|
Molecular Formula | Zn |
Molecular Weight (g/mol) | 65.38 |
MDL Number | MFCD00011291 |
InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
Synonym | dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust |
PubChem CID | 23994 |
ChEBI | CHEBI:30185 |
IUPAC Name | zinc |
SMILES | [Zn] |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10571852
|
Thermo Scientific Acros
367260100 |
10 g | Glass bottle |
£112.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10068681
|
Thermo Scientific Acros
367260500 |
50 g | Glass bottle |
£381.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis, Reduction reagentChemische Identifikatoren
7440-66-6 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
23994 | |
zinc |
Zn | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
CHEBI:30185 | |
[Zn] |
Spezifikation
Gray | |
420.0°C | |
7.14 | |
Zn | |
MFCD00011291 | |
15, 10326 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
65.38 | |
Trace Metal Basis | |
Zinc, 99.999% |
7440-66-6 | |
907.0°C | |
100.00% | |
Zn | |
01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
99.999% | |
Powder |
Sicherheit und Handhabung
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.
Catches fire spontaneously if exposed to air.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Zinc, powder
Signalwort
- Danger
Gefahrenkategorie
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric solid Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Gefahrenhinweis
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P273-Avoid release to the environment.
RUO – Research Use Only