missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis, Reduction reagent
Specifications
Specifications
| Melting Point | 420.0°C |
| Boiling Point | 907.0°C |
| Density | 7.14g/mL |
| Quantity | 50 g |
| Packaging | Glass bottle |
| Assay Percent Range | 100.00% |
| Form | Powder |
| Linear Formula | Zn |
| Fieser | 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 |
| Merck Index | 15, 10326 |
| Show More |
Safety and Handling
Product Identifier
- Zinc, powder
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use sand to extinguish.
- MIXTURE LIST-Contain: Zinc
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?