missing translation for 'onlineSavingsMsg'
Learn More

Zinc, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals

Product Code. p-3734564 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10786062

Brand: Thermo Scientific Acros 194500050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Melting Point 420.0°C
Boiling Point 907.0°C
Density 7.14g/mL
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 100.00%
Form Powder
Linear Formula Zn
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Merck Index 15, 10326
Formula Weight 65.38
Percent Purity 99.995%
Grade Trace Metal Basis
Chemical Name or Material Zinc
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Zinc, powder
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use sand to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Zinc

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.