missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether

Product Code. 11358378
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358378 100 g 100g
11348378 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358378 Supplier Thermo Scientific Alfa Aesar Supplier No. 039560.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]

Specifications

Quantity 100 g
Solubility Information Immiscible with cold water.
Formula Weight 351.79
Physical Form Liquid
Chemical Name or Material Zinc 2-ethylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.