missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541198
This item is not returnable.
View return policy
Description
Peptide sequence: CGKAFFQKSN LHSHQKTHSG ERTYRCSECG KTFIRKLNLS LHKKTHTGQK Sequence homology: Human: 100%.
ZIM3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 11 C2H2-type zinc fingers and 1 KRAB domain. ZIM3 may be involved in transcriptional regulation.
Specifications
| ZIM3 | |
| Polyclonal | |
| Unconjugated | |
| ZIM3 | |
| ZIM3; Zinc finger imprinted 3; zinc finger protein 657; zinc finger, imprinted 3; ZNF657 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96PE6 | |
| ZIM3 | |
| Synthetic peptide directed towards the middle region of human ZIM3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction