missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ZIM3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZIM3 Polyclonal specifically detects ZIM3 in Human samples. It is validated for Western Blot.Specifications
| ZIM3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MGC138876, MGC138877, zinc finger imprinted 3, zinc finger protein 657, zinc finger, imprinted 3, ZNF657 | |
| ZIM3 | |
| IgG | |
| 54 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_443114 | |
| 114026 | |
| Synthetic peptide directed towards the middle region of human ZIM3. Peptide sequence CGKAFFQKSNLHSHQKTHSGERTYRCSECGKTFIRKLNLSLHKKTHTGQK. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title