missing translation for 'onlineSavingsMsg'
Learn More

Zilpaterol, TRC

Product Code. 30041246
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041246 10 mg 10mg
30041211 2.5 mg 2.5mg
30040712 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041246 Supplier Toronto Research Chemicals Supplier No. TRCZ43000010MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119520-05-7
Molecular Formula C14 H19 N3 O2
Molecular Weight (g/mol) 261.32
Synonym Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 4,5,6,7-tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]-, (6R,7R)-rel-Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 4,5,6,7-tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]-, trans-(±)-, RU 42173, Zilpaterol, trans-(±)-Zilpaterol
SMILES CC(C)N[C@@H]1CCN2C(=O)Nc3cccc([C@H]1O)c23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H19N3O2/c1-8(2)15-11-6-7-17-12-9(13(11)18)4-3-5-10(12)16-14(17)19/h3-5,8,11,13,15,18H,6-7H2,1-2H3,(H,16,19)/t11-,13-/m1/s1
Formula Weight 261.1477
Percent Purity >95%
Chemical Name or Material Zilpaterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.