missing translation for 'onlineSavingsMsg'
Learn More

Zilpaterol, Dr. Ehrenstorfer

Product Code. 30309595
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309595 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309595 Supplier Dr. Ehrenstorfer Supplier No. DREC17949000
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zilpaterol
CAS 119520-05-7
Analyte or Component Names Zilpaterol
Shipping Condition +20°C
Molecular Formula C14 H19 N3 O2
Certifications/Compliance ISO/IEC 17025
Synonym Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 4,5,6,7-tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]-, (6R,7R)-rel-Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 4,5,6,7-tetrahydro-7-hydroxy-6-[(1-methylethyl)amino]-, trans-(±)-, RU 42173, Zilpaterol, trans-(±)-Zilpaterol
InChI Formula InChI=1S/C14H19N3O2/c1-8(2)15-11-6-7-17-12-9(13(11)18)4-3-5-10(12)16-14(17)19/h3-5,8,11,13,15,18H,6-7H2,1-2H3,(H,16,19)/t11-,13-/m1/s1
SMILES CC(C)N[C@@H]1CCN2C(=O)Nc3cccc([C@H]1O)c23
Molecular Weight (g/mol) 261.32
Quantity 10 mg
Formula Weight 261.1477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.