missing translation for 'onlineSavingsMsg'
Learn More

Zileuton, MedChemExpress

Product Code. p-200097980
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16482117

Brand: Medchem Express HY14164

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zileuton is a potent and selective inhibitor of 5-lipoxygenase with antiasthmatic properties.

Zileuton is a potent and selective inhibitor of 5-lipoxygenase with antiasthmatic properties.

Specifications

Chemical Name or Material Zileuton
Physical Form Solid
CAS 111406-87-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H12N2O2S
For Use With (Application) COVID-19-immunoregulation
Synonym A 64077 Abbott 64077
Solubility Information DMSO : ≥ 100 mg/mL (423.21 mM)
SMILES CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1
Molecular Weight (g/mol) 236.29
Formula Weight 236.29
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.