missing translation for 'onlineSavingsMsg'
Learn More

Zileuton, MedChemExpress

Product Code. 16482117
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482117 50 mg 50mg
16425978 10 mg 10mg
16406406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482117 Supplier Medchem Express Supplier No. HY14164
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zileuton is a potent and selective inhibitor of 5-lipoxygenase with antiasthmatic properties.

Zileuton is a potent and selective inhibitor of 5-lipoxygenase with antiasthmatic properties.

Specifications

Chemical Name or Material Zileuton
Physical Form Solid
CAS 111406-87-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H12N2O2S
For Use With (Application) COVID-19-immunoregulation
Synonym A 64077 Abbott 64077
Solubility Information DMSO : ≥ 100 mg/mL (423.21 mM)
SMILES CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1
Molecular Weight (g/mol) 236.29
Formula Weight 236.29
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.