missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus VLP Mouse anti-Virus, Clone: IG3-1-A12-G9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14850-0.5MG
This item is not returnable.
View return policy
Description
Zika Virus VLP Monoclonal antibody specifically detects Zika Virus VLP in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Zika Virus VLP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| polyprotein, partial | |
| Recombinant Zika virus VLPs (comprising envelope, pre-membrane and membrane proteins), produced in HEK293 cells | |
| 0.5 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| Purified |
| Western Blot, ELISA | |
| IG3-1-A12-G9 | |
| Unconjugated | |
| PBS pH 7.4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction