missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus VLP Mouse anti-Virus, Clone: ID5-2-H7-G3, Novus Biologicals™
Mouse Monoclonal Antibody
£409.00 - £1055.00
Specifications
| Antigen | Zika Virus VLP |
|---|---|
| Clone | ID5-2-H7-G3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18347134
|
Novus Biologicals
NBP3-14851-0.1MG |
0.1 mg |
£409.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18339266
|
Novus Biologicals
NBP3-14851-0.5MG |
0.5 mg |
£1055.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zika Virus VLP Monoclonal antibody specifically detects Zika Virus VLP in Virus samples. It is validated for Western Blot, ELISASpecifications
| Zika Virus VLP | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| polyprotein, partial | |
| IgG1 κ | |
| Protein G purified | |
| Antibody |
| ID5-2-H7-G3 | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS pH 7.4 | |
| Recombinant Zika virus VLPs (comprising envelope, pre-membrane and membrane proteins), produced in HEK293 cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title