missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) Envelope Antibody (35), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05856-50ul
This item is not returnable.
View return policy
Description
Zika Virus (SPH2015) Envelope Monoclonal antibody specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Sandwich ELISA Capture
Specifications
| Zika Virus (SPH2015) Envelope | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Sandwich ELISA Capture | |
| 35 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Zika Envelope | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val593-Lys699). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction