missing translation for 'onlineSavingsMsg'
Learn More

Zika Virus (SPH2015) Envelope Antibody (09), DyLight 350 , Novus Biologicals™

Product Code. 18417289 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417289 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417289 Supplier Novus Biologicals Supplier No. NBP305849UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zika Virus (SPH2015) Envelope Monoclonal antibody specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus (SPH2015) Envelope
Applications Western Blot, ELISA
Classification Monoclonal
Clone 09
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Zika Envelope
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/ Envelope Protein (ALU33341.1; Val593-Lys699).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.