missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) Envelope Antibody (09), Alexa Fluor™ 750 , Novus Biologicals™
Shop All Bio Techne ProductsDescription
Zika Virus (SPH2015) Envelope Monoclonal antibody specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Zika Virus (SPH2015) Envelope |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 09 |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Zika Envelope |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/ Envelope Protein (ALU33341.1; Val593-Lys699). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?