missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zika Virus NS4B (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17830956
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17830956 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17830956 Supplier Invitrogen™ Supplier No. PA5143382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zika virus NS4B-transfected 293T, Zika virus NS4B-infected C6/36.

Zika virus (ZIKV) NS4B protein is a membranotropic multifunctional protein Despite its versatile functioning, its topology and dynamics are not entirely understood There is no X-ray or cryo-EM structure available for any flaviviral NS4B full-length protein [PMID: 36037701].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS4B (strain H/PF/2013)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias Zika virus Non-structural protein 4B; Zika virus NS4B; ZIKV Non-structural protein 4B; ZIKV NS4B
Host Species Rabbit
Immunogen Full length Zika virus NS4B recombinant protein. (Zika virus (strain H/PF/2013)).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.