missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Zika virus NS4A Polyclonal antibody specifically detects Zika virus NS4A in Virus samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Zika virus NS4A |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | Non-structural protein 4A, ZIKV NS4A |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of Zika virus NS4A (Zika virus (strain H/PF/2013)). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?