missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zika Virus NS1 (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (201)

Rabbit Recombinant Monoclonal Antibody
£132.00 - £357.00
Specifications
| Antigen | Zika Virus NS1 (strain Zika SPH2015) |
|---|---|
| Clone | 201 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534108
![]() |
Invitrogen™
MA560381 |
100 μL |
£357.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30533784
|
Invitrogen™
MA560382 |
20 μL |
£132.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zika Virus NS1 (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. & Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.Specifications
| Zika Virus NS1 (strain Zika SPH2015) | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 protein (Val796-Leu1157). | |
| Antibody |
| 201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| Zika NS1; ZIKV NS1; ZIKV-NS1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title