missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Zika Virus NS1 Monoclonal Antibody (E107)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524585
This item is not returnable.
View return policy
Description
MA5-24585 does not cross react with the following viruses: Dengue 1-4, TBE virus, West Nile virus, Yellow Fever virus, Japanese Encephalitis virus and Chikungunya virus.
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. & Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
Specifications
| Zika Virus NS1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| E107 | |
| Unconjugated | |
| Zika NS1; ZIKV NS1; ZIKV-NS1 | |
| recombinant Zika Virus NS1 protien. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction