missing translation for 'onlineSavingsMsg'
Learn More

Zika Virus NS1 Human anti-Virus, Clone: B4, Novus Biologicals™

Product Code. 18345866 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
0.5 mg
Unit Size:
0.1mg
0.25mg
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345866 0.25 mg 0.25mg
18326864 0.1 mg 0.1mg
18309764 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18345866 Supplier Novus Biologicals Supplier No. NBP3148390.25MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Zika Virus NS1 Monoclonal antibody specifically detects Zika Virus NS1 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS1
Applications ELISA
Classification Monoclonal
Clone B4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS pH 7.4
Gene Alias polyprotein, partial
Host Species Human
Immunogen Full length recombinant NS1 protein of Zika virus produced in HEK293 cells
Purification Method Protein G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.