missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Zika Virus IgG |
| Applications | Western Blot, ELISA |
| Dilution | Western Blot, ELISA |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?