missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Zika Virus IgG |
| Applications | ELISA |
| Classification | Polyclonal |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Immunogen | Zika Capsid antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Capsid. Amino Acid Squence: RIINARKEKKRRGADTS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?