missing translation for 'onlineSavingsMsg'
Learn More

Zika virus (H/PF/2013) NS1 Antibody (990948) [DyLight 755], Novus Biologicals™

Product Code. 30762593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762593 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762593 Supplier Novus Biologicals Supplier No. FAB9920Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zika virus (H/PF/2013) NS1 Monoclonal antibody specifically detects Zika virus (H/PF/2013) NS1 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika virus (H/PF/2013) NS1
Applications ELISA
Classification Monoclonal
Clone 990948
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived recombinant Viral Zika Virus (H/PF/2013) NS1, Val796 & Ser799-Ser1148, with a C-terminal 6-His tag , Accession # AHZ13508
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.