missing translation for 'onlineSavingsMsg'
Learn More

Zidovudine, MedChemExpress

Product Code. 16437296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437296 100 mg 100mg
16421856 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437296 Supplier Medchem Express Supplier No. HY17413
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zidovudine is a nucleoside reverse transcriptase inhibitor (NRTI), widely used to treat HIV infection. Zidovudine increases CRISPR/Cas9-mediated editing frequency.

Zidovudine is a nucleoside reverse transcriptase inhibitor (NRTI), widely used to treat HIV infection. Zidovudine increases CRISPR/Cas9-mediated editing frequency.

Specifications

Chemical Name or Material Zidovudine
Physical Form Solid
CAS 30516-87-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H13N5O4
For Use With (Application) COVID-19-anti-virus
Synonym Azidothymidine AZT ZDV
Solubility Information DMSO : ≥ 100 mg/mL (374.20 mM) ∣H2O : 16.67 mg/mL (62.38 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C(C)=CN1[C@@H](C2)O[C@@H]([C@H]2N=[N+]=[N-])CO)NC1=O
Molecular Weight (g/mol) 267.24
Formula Weight 267.24
Percent Purity 99.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.