missing translation for 'onlineSavingsMsg'
Learn More

ZIC3 Antibody [PerCP], Novus Biologicals™

Product Code. 30734567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734567 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734567 Supplier Novus Biologicals Supplier No. AF5310C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ZIC3 Polyclonal antibody specifically detects ZIC3 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias heterotaxy 1, HTXHTX1, Zic family member 3 (odd-paired homolog, Drosophila), Zinc finger protein 203, zinc finger protein ZIC 3, ZNF203Zinc finger protein of the cerebellum 3
Host Species Sheep
Immunogen E. coli-derived recombinant human ZIC3, His380-Val467, Accession # O60481
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7547
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.