missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC2 Polyclonal Antibody

Brand: Thermo Scientific PA5113616
This item is not returnable.
View return policy
Description
Specifications
| ZIC2 | |
| Polyclonal | |
| Unconjugated | |
| ZIC2 | |
| cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 2; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; zinc finger protein ZIC 2; zinc finger protein Zic2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7546 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95409 | |
| ZIC2 | |
| Synthetic peptide directed towards the C terminal region of human ZIC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction