missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIC1/ZIC2/ZIC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599208
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZIC1/2/3.
Zic1 encodes a zinc finger protein expressed in the developing or matured central nervous system in a highly restricted manner. Zic is expressed in granule cells that make synaptic contact with Purkinje cells. Zic1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation, becoming restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. The Zic1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic1 gene contains a region-specific enhancer determined to be essential in in-vivo and in-vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family. Zic2 and Zic3 are highly similar genes. Zic2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation.
Specifications
| ZIC1/ZIC2/ZIC3 | |
| Polyclonal | |
| Unconjugated | |
| ZIC3 | |
| Bn; CRS6; GENA 29; heterotaxy 1; HPE5; HTX; HTX1; Ka; Ku; kumba; odd-paired homolog; RGD1561261; VACTERLX; Zic; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); zic protein member 1; Zic1; Zic2; Zic3; Zinc finger protein 201; zinc finger protein 203; Zinc finger protein of the cerebellum 1; zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; zinc finger protein ZIC 1; Zinc finger protein ZIC 2; zinc finger protein ZIC 3; ZNF201; ZNF203 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22771, 22772, 22773, 7545, 7546, 7547 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60481, O95409, P46684, Q15915, Q62520, Q62521 | |
| ZIC1, ZIC2, ZIC3 | |
| A synthesized peptide derived from human ZIC1(Accession Q15915), corresponding to amino acid residues G328-R378. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction