missing translation for 'onlineSavingsMsg'
Learn More

ZIC1 Rabbit anti-Human, Mouse, Rat, Clone: 6B5O9, Novus Biologicals™

Product Code. 18370256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370256 20 μg 20µL
18364433 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370256 Supplier Novus Biologicals Supplier No. NBP31577420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ZIC1 Monoclonal antibody specifically detects ZIC1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC1
Applications Western Blot
Classification Monoclonal
Clone 6B5O9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Zic family member 1 (odd-paired homolog, Drosophila), ZICZic family member 1 (odd-paired Drosophila homolog), Zinc finger protein 201, Zinc finger protein of the cerebellum 1, ZNF201zinc finger protein ZIC 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ZIC1 (Q15915).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7545
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.