missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-488F
This item is not returnable.
View return policy
Description
ZIC1 Polyclonal specifically detects ZIC1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| ZIC1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Q15915 | |
| ZIC1 | |
| A synthetic peptide made to an internal portion of the human Zic1 protein (between residues 20-75) [UniProt Q15915] | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 7545 | |
| Human, Mouse | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Zic family member 1 (odd-paired homolog, Drosophila), ZICZic family member 1 (odd-paired Drosophila homolog), Zinc finger protein 201, Zinc finger protein of the cerebellum 1, ZNF201zinc finger protein ZIC 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Predicted to react with rat based on 100% sequence homology. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction