missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC Pan Polyclonal Antibody, Invitrogen™
Description
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigen | ZIC Pan |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZIC2 |
| Gene Accession No. | O60481, O95409, Q15915, Q8N9L1, Q96T25, P46684, Q61467, Q62520, Q62521, Q7TQ40 |
| Gene Symbols | ZIC1, ZIC2, ZIC3, ZIC4, ZIC5 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?