missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIC Pan Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103289
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZIC1/2/3/4/5.
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq, Jul 2008].
Specifications
| ZIC Pan | |
| Polyclonal | |
| Unconjugated | |
| ZIC3 | |
| 1700049L20Rik; Bn; CRS6; GENA 29; heterotaxy 1; HPE5; HTX; HTX1; Ka; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 5; Odd paired-related protein; odd-paired homolog; odd-paired related; Opa-related protein; Opr; RGD1561261; VACTERLX; Zic; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); Zic family member 4; Zic family member 4 (odd-paired homolog, Drosophila); Zic family member 5; zic family member 5 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; ZIC2; Zic3; ZIC4; ZIC5; zinc family member 4 protein HZIC4; zinc family member 5 protein; zinc finger protein 201; Zinc finger protein 203; zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; zinc finger protein of the cerebellum 3; Zinc finger protein of the cerebellum 4; zinc finger protein of the cerebellum 5; Zinc finger protein ZIC 1; zinc finger protein ZIC 2; Zinc finger protein ZIC 3; zinc finger protein ZIC 4; Zinc finger protein ZIC 5; zinc finger protein ZIC 5; LOW QUALITY PROTEIN: zinc finger protein ZIC 5; zinc finger protein Zic5; ZNF201; ZNF203 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22771, 22772, 22773, 22774, 315882, 361095, 361096, 367944, 64618, 65100, 7545, 7546, 7547, 84107, 85416 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60481, O95409, P46684, Q15915, Q61467, Q62520, Q62521, Q7TQ40, Q8N9L1, Q96T25 | |
| ZIC1, ZIC2, ZIC3, ZIC4, Zic5 | |
| A synthesized peptide derived from human ZIC1(Accession Q15915), corresponding to amino acid residues G328-R378. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction