missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Mouse anti-Human, Clone: PCRP-ZHX3-1D11, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | ZHX3 |
|---|---|
| Clone | PCRP-ZHX3-1D11 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18354084
|
Novus Biologicals
NBP3-13888-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373844
|
Novus Biologicals
NBP3-13888-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZHX3 Monoclonal antibody specifically detects ZHX3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Protein ArraySpecifications
| ZHX3 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H4I2 | |
| 23051 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-ZHX3-1D11 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
| Recombinant full-length human ZHX3 protein (Uniprot: Q9H4I2) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title