missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZHX3 Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-68219PCP
This item is not returnable.
View return policy
Description
ZHX3 Polyclonal antibody specifically detects ZHX3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| ZHX3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23051 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| PerCP | |
| FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human zinc fingers and homeoboxes 3 using the numbering given in entry NP_055850.1 (GeneID 23051). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction