missing translation for 'onlineSavingsMsg'
Learn More

ZHX3 Antibody, Novus Biologicals™

Product Code. 18386678 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386678 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386678 Supplier Novus Biologicals Supplier No. H00023051B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ZHX3 Polyclonal antibody specifically detects ZHX3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZHX3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. AAH68569.1
Gene Alias FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1
Host Species Mouse
Immunogen ZHX3 (AAH68569.1, 1 a.a. - 969 a.a.) full-length human protein. MASKRKSTTPCMIPVKTVVLQDASMEAQPAETLPEGPQQDLPPEASAASSEAAQNPSSTDGSTLANGHRSTLDGYLYSCKYCDFRSHDMTQFVGHMNSEHTDFNKDPTFVCSGCSFLAKTPEGLSLHNATCHSGEASFVWNVAKPDNHVVVEQSIPESTSTPDLAGEPSAEGADGQAEIIITKTPIMKIMKGKAEAKKIHTLKENVPSQPVGEALPKLSTGEMEVREGDHSFINGAVPVSQASASSAKNPHAANGPLIGTVPVLPAGIAQFLSLQQQPPVHAQHHVHQPLPTAKALPKVMIPLSSIPTYNAAMDSNSFLKNSFHKFPYPTKAELCYLTVVTKYPEEQLKIWFTAQRLKQGISWSPEEIEDARKKMFNTVIQSVPQPTITVLNTPLVASAGNVQHLIQAALPGHVVGQPEGTGGGLLVTQPLMANGLQATSSPLPLTVTSVPKQPGVAPINTVCSNTTSAVKVVNAAQSLLTACPSITSQAFLDASIYKNKKSHEQLSALKGSFCRNQFPGQSEVEHLTKVTGLSTREVRKWFSDRRYHCRNLKGSRAMIPGDHSSIIIDSVPEVSFSPSSKVPEVTCIPTTATLATHPSAKRQSWHQTPDFTPTKYKERAPEQLRALESSFAQNPLPLDEEMDRLRSETKMTRREIDSWFSERRKKVNAEETKKAEENASQEEEEAAEDEGGEEDLASELRVSGENGSLEMPSSHILAERKVSPIKINLKNLRVTEANGRNEIPGLGACDPEDDESNKLAEQLPGKVSCKKTAQQRHLLRQLFVQTQWPSNQDYDSIMAQTGLPRPEVVRWFGDSRYALKNGQLKWYEDYKRGNFPPGLLVIAPGNRELLQDYYMTHKMLYEEDLQNLCDKTQMSSQQKQTEFDLINVKDWPVWETACHVEEPNPTLCCHMPFPCPAAGHLGELPESSQTAQSLPLPSACPPPSKQQARWGSHQFFLPQCRTFPLPSNG
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23051
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.