missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZHX2 Recombinant Rabbit Monoclonal Antibody (23GB4270), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

£179.00 - £485.00

Specifications

Antigen ZHX2
Clone 23GB4270
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088956
View Documents
Invitrogen™
MA553724
100 μL
£485.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088965
View Documents
Invitrogen™
MA553725
20 μL
£179.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZHX2 Recombinant Monoclonal antibody specifically detects ZHX2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot
Specifications

Specifications

ZHX2
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q80VX4, Q8C0C0, Q9Y6X8
22882, 314988, 387609
IgG
Affinity chromatography
Antibody
23GB4270
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AFP regulator 1; AFR1; Afr-1; alpha fetoprotein regulation 1, adult; alpha-fetoprotein regulator 1; KIAA0854; LOW QUALITY PROTEIN: zinc fingers and homeoboxes protein 2; mKIAA0854; Raf; Regulator of AFP; transcription factor ZHX2; Zhx2; Zinc finger and homeodomain protein 2; zinc fingers and homeoboxes 2; zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2
A synthesized peptide derived from human ZHX2 (260-500AA).
Primary
-20°C
ZHX2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.