missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20136-1-AP-20UL
This item is not returnable.
View return policy
Description
Zinc-fingers and homeobox (ZHX) proteins are transcription factors that interact with the activation domain of the A subunit of nuclear factor-Y (NF-YA). ZHX1-3 are ubiquitously expressed proteins expressed in various tissues. They act as transcriptional repressors and localize to the nucleus. The ZHX proteins contain two Cys(2)-His(2)-type zinc-finger motifs and five homeodomains (HDs). These domains allow the ZHX proteins to form homodimers, but they can also form heterodimers with each other. However, this dimerization is not required for repressor activity. Hypermethylation-mediated silencing of ZHX2 is an epigenetic event involved in hepatocellular carcinoma (HCC).Specifications
| ZHX2 | |
| Polyclonal | |
| Unconjugated | |
| ZHX2 | |
| AFP regulator 1, AFR1, Alpha fetoprotein regulator 1, KIAA0854, RAF, Regulator of AFP, ZHX2, zinc fingers and homeoboxes 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22882, 314988, 387609 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80VX4, Q8C0C0, Q9Y6X8 | |
| Zhx2 | |
| ZHX2 Fusion Protein Ag14088 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction