missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZGPAT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23203-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcription repressor that specifically binds the 5′-GGAG[GA]A[GA]A-3′ consensus sequence. Represses transcription by recruiting the chromatin multiprotein complex NuRD to target promoters. Negatively regulates expression of EGFR, a gene involved in cell proliferation, survival and migration. Its ability to repress genes of the EGFR pathway suggest it may act as a tumor suppressor. Able to suppress breast carcinogenesis. Isoform 4: Antagonizes the transcription repression by isoform 1 by competing for the binding of the NuRD complex. Does not bind DNA.Specifications
| ZGPAT | |
| Polyclonal | |
| Unconjugated | |
| ZGPAT | |
| GPATC6, GPATCH6, KIAA1847, ZC3H9, ZC3HDC9, ZGPAT, ZIP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229007, 84619 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N5A5, Q8VDM1 | |
| ZGPAT | |
| ZGPAT Fusion Protein Ag19620 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction