missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZGPAT Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | ZGPAT |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662516
|
Novus Biologicals
NBP2-41146-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122589
|
Novus Biologicals
NBP2-41146 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ZGPAT | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 84619 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZGPAT. The immunogen is located within the last 50 amino acids of ZGPAT. Amino Acid Squence: CQEQRKADTHKKMTEF | |
| Primary | |
| Multiple isoforms of ZGPAT are known to exist. |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Human | |
| GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein | |
| ZGPAT | |
| IgY | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title