missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZG16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562545
This item is not returnable.
View return policy
Description
Immunogen sequence: QVSGKYKWYL KKLVFVTDKG RYLSFGKDSG TSFNAVPLHP NTVLRFISGR SGSLIDAIGL HWDVYPTS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.
Specifications
| ZG16 | |
| Polyclonal | |
| Unconjugated | |
| ZG16 | |
| 1810010M01Rik; AI593689; jacalin-like lectin domain containing; JCLN; JCLN1; Secretory lectin ZG16; ZG16; ZG16A; Zg16p; Zg-16p; Zymogen granule membrane protein 16; zymogen granule protein 16; zymogen granule protein 16 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 653808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60844 | |
| ZG16 | |
| Recombinant protein corresponding to Human ZG16. Recombinant protein control fragment (Product #RP-99618). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction