missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZG16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17397-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| ZG16 | |
| Polyclonal | |
| Unconjugated | |
| ZG16 | |
| hZG16, JCLN, JCLN1, Secretory lectin ZG16, ZG16, ZG16A, Zymogen granule protein 16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171449, 653808, 69036 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60844, Q8CJD3, Q8K0C5 | |
| Zg16 | |
| ZG16 Fusion Protein Ag11332 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction