missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE9, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00009372-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MENYFQAEAYNLDKVLDEFEQNEDETVSSTLLDTKWNKILDPPSHRLSFNPTLASVNESAVSNESQPQLKVFSLAHSAPLTTEEEDHCANGQDCNLNPEIATMWIDENAVAEDQLIKRNYSWDDQCSAVEVGEKKCGNLACLPDEKNVLVVAVMHNCDKRTLQNDLQDCNNYNSQSLMDAFSCSLDNENRQTDQFSFSINESTEKDMNSEKQMDPLNRPKTEGRSVNHLCPTSSDSLASVCSPSQLKDDGSIGRDPSMSAITSLTVDSVISSQGTDGCPAVKKQENYIPDEDLTGKISSPRTDLGSPNSFSHMSEGILMKKEPAEESTTEESLRSGLPLLLKPDMPNGSGRNNDCERCSDCLVPNEVRADENEGYEHEETLGTTEFLNMTEHFSESQDMTNWKLTKLNEMNDSQVNEEKEKFLQISQPEDTNGDSGGQCVGLADAGLDLKGTCISESEECDFSTVIDTPAANYLSNGCDSYGMQDPGVSFVPKTLPSKEDSVTEEKEIEESKSECYSNIYEQRGNEATEGSGLLLNSTGDLMKKNYLHNFCSQVPSVLGQSSPKVVASLPSISVPFGGARPKQPSNLKLQIPKPLSDHLQNDFPANSGNNTKNKNDILGKAKLGENSATNVCSPSLGNISNVDTNGEHLESYEAEISTRPCLALAPDSPDNDLRAGQFGISARKPFTTLGEVAPVWVPDSQAPNCMKCEARFTFTKRRHHCRACGKVFCASCCSLKCKLLYMDRKEARVCVICHSVLMNGKYSpecifications
| ZFYVE9 | |
| Polyclonal | |
| No additive | |
| NM_007323 | |
| ZFYVE9 | |
| ZFYVE9 (NP_015562.1, 1 a.a. ∼ 762 a.a) full-length human protein. | |
| RUO | |
| 9372 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| zinc finger, FYVE domain containing 9 | |
| MADHIP/NSP/SARA/SMADIP | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction