missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFYVE9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106937
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZFYVE9.
Early endosomal protein that functions to recruit SMAD2/SMAD3 to intracellular membranes and to the TGF-beta receptor. Plays a significant role in TGF-mediated signaling by regulating the subcellular location of SMAD2 and SMAD3 and modulating the transcriptional activity of the SMAD3/SMAD4 complex. Possibly associated with TGF-beta receptor internalization.
Specifications
| ZFYVE9 | |
| Polyclonal | |
| Unconjugated | |
| Zfyve9 | |
| E030027L17; hSARA; MAD, mothers against decapentaplegic homolog interacting protein, receptor activation anchor; MADH-interacting protein; MADHIP; Mothers against decapentaplegic homolog-interacting protein; Novel serine protease; NSP; PPP1R173; protein phosphatase 1, regulatory subunit 173; Receptor activation anchor; SARA; Smad anchor for receptor activation; SMADIP; ZFYVE9; Zinc finger FYVE domain-containing protein 9; zinc finger FYVE-type containing 9; zinc finger, FYVE domain containing 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 230597, 9372 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95405 | |
| Zfyve9 | |
| A synthesized peptide derived from human ZFYVE9(Accession O95405), corresponding to amino acid residues Y1302-K1352. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction