missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE28 Mouse anti-Human, Clone: LST2/2426, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | ZFYVE28 |
|---|---|
| Clone | LST2/2426 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
ZFYVE28 Monoclonal specifically detects ZFYVE28 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| ZFYVE28 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| hLst2, KIAA1643, lateral signaling target protein 2 homolog, LST2, LYST2, MGC43699, MGC61592, Zinc finger FYVE domain-containing protein 28, zinc finger, FYVE domain containing 28 | |
| Recombinant full-length human ZFYVE28 protein | |
| Primary | |
| Store at 4°C. |
| LST2/2426 | |
| Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 57732 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title