Learn More
Invitrogen™ ZFYVE27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590842
Description
Immunogen sequence: VEFFRVVSEY RASLQQRMNP KQEEHAFESP PPPDVGGKDG LMDSTPALTP TEDLTPGSVE EAEEAEPDEE FKDAIEETHL VVLEDDEGAP CPAEDELALQ DNGFLSKNEV LRSKVSRLTE RLRKRYPTNN FGNCTGCSAT FSVLKKRRSC SNCGNSFCSR CCSFKVPKSS MGATAPEAQR ETVFVCASCN QTLSK; Positive Samples: Rat eye; Cellular Location: Cell membrane, Cell projection, Endoplasmic reticulum membrane, Multi-pass membrane protein, Recycling endosome membrane, growth cone membrane.
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
Specifications
| ZFYVE27 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE27 | |
| 2210011N02Rik; 9530077C24Rik; AI426636; AI593546; AI835681; im:7137347; LOW QUALITY PROTEIN: protrudin; PROTRUDIN; RP11-459F3.2; Spastic paraplegia 33 protein; SPG33; zfyve27; zgc:153156; zinc finger FYVE domain containing 27; zinc finger FYVE domain-containing protein 27; zinc finger FYVE-type containing 27; zinc finger, FYVE domain containing 27; zinc finger, FYVE domain containing 27 isoform c | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 118813, 309376 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.22 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5T4F4, Q6P7B7 | |
| ZFYVE27 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 217-411 of human ZFYVE27 (NP_6531893). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.